missing translation for 'onlineSavingsMsg'
Learn More

8-Anilinonaphthalene-1-sulfonic acid ammonium salt, 98%

Catalog No. p-7043102
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6553806 5 g
AAJ6553814 25 g
AAJ6553822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6553806 Supplier Thermo Scientific Chemicals Supplier No. J6553806
Only null left

Fluorescent probe for proteins on polyacrylamide gels | CAS: 28836-03-5 | C16H16N2O3S | 316.38 g/mol

ANS forms an inclusion complex with cyclodextrin. Such model systems are useful to mimic biological recognition and can be studied by measuring the change in fluorescence of free-ANS to complexed-ANS. When ANS enters the hydrophobic core of cyclodestrin, it’s fluorescence increases. It is commonly used to study molecular assemblies of surfactants and amphiphilic polymers because a blue shift of the emission maximum indicates the fluorophore is located in less polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ANS forms an inclusion complex with cyclodextrin. Such model systems are useful to mimic biological recognition and can be studied by measuring the change in fluorescence of free-ANS to complexed-ANS. When ANS enters the hydrophobic core of cyclodestrin, it’s fluorescence increases. It is commonly used to study molecular assemblies of surfactants and amphiphilic polymers because a blue shift of the emission maximum indicates the fluorophore is located in less polar media.

Solubility
Soluble in water, 1N NaOH, and methanol.

Notes
Light & Air Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 28836-03-5
Molecular Formula C16H16N2O3S
Molecular Weight (g/mol) 316.38
MDL Number MFCD00012560
InChI Key IPBNQYLKHUNLQE-UHFFFAOYSA-N
Synonym ammonium 8-phenylamino naphthalene-1-sulfonate, 8-anilino-1-naphthalenesulfonic acid ammonium salt, unii-gt7mn8dkd7, gt7mn8dkd7, ammonium 8-anilino-1-naphthalenesulfonate, 1-naphthalenesulfonic acid, 8-phenylamino-, monoammonium salt, 1-naphthalenesulfonic acid, 8-phenylamino-, ammonium salt 1:1, ansa, 1-anilinonaphthalene-8-sulfonic acid ammonium salt, 82-76-8 parent
PubChem CID 120066
IUPAC Name azanium;8-anilinonaphthalene-1-sulfonate
SMILES [NH4+].[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1

Specifications

Melting Point 242°C to 244°C
Color Yellow to Green
Quantity 5 g
Beilstein 3581235
Sensitivity Air and light sensitive; Hygroscopic
Solubility Information Soluble in water,1N NaOH,and methanol.
Formula Weight 316.37
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 8-Anilinonaphthalene-1-sulfonic acid ammonium salt
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 249-265-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only