Learn More
8-Anilinonaphthalene-1-sulfonic acid, 95%
Fluorescent probe for proteins on polyacrylamide gels | CAS: 82-76-8 | C16H13NO3S | 299.344 g/mol
$85.26 - $686.70
Chemical Identifiers
| CAS | 82-76-8 |
|---|---|
| Molecular Formula | C16H13NO3S |
| Molecular Weight (g/mol) | 299.344 |
| MDL Number | MFCD00003998 |
| InChI Key | FWEOQOXTVHGIFQ-UHFFFAOYSA-N |
| Synonym | 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid |
| PubChem CID | 1369 |
| ChEBI | CHEBI:39708 |
| IUPAC Name | 8-anilinonaphthalene-1-sulfonic acid |
| SMILES | C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6533206
|
Thermo Scientific Chemicals
J6533206 |
5 g |
Each for $85.26
|
|
|||||
|
AAJ6533214
|
Thermo Scientific Chemicals
J6533214 |
25 g |
Each for $194.18
|
|
|||||
|
AAJ6533222
|
Thermo Scientific Chemicals
J6533222 |
100 g |
Each for $686.70
|
|
|||||
Description
Fluorescent probe for proteins on polyacrylamide gels
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 82-76-8 | |
| 299.344 | |
| FWEOQOXTVHGIFQ-UHFFFAOYSA-N | |
| 1369 | |
| 8-anilinonaphthalene-1-sulfonic acid |
| C16H13NO3S | |
| MFCD00003998 | |
| 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid | |
| CHEBI:39708 | |
| C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O |
Specifications
| 82-76-8 | |
| Black to Gray | |
| MFCD00003998 | |
| 1843887 | |
| 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid | |
| C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O | |
| 299.344 | |
| CHEBI:39708 | |
| 95% | |
| 8-Anilinonaphthalene-1-sulfonic acid |
| 215°C to 217°C | |
| C16H13NO3S | |
| 5 g | |
| 14,661 | |
| FWEOQOXTVHGIFQ-UHFFFAOYSA-N | |
| 8-anilinonaphthalene-1-sulfonic acid | |
| 1369 | |
| 299.34 | |
| Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-438-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only