missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Anilino-1-naphthalenesulfonic Acid 95.0+%, TCI America™
$150.98 - $150.98
Chemical Identifiers
| CAS | 82-76-8 |
|---|---|
| Molecular Formula | C16H13NO3S |
| Molecular Weight (g/mol) | 299.344 |
| MDL Number | MFCD00003998 |
| InChI Key | FWEOQOXTVHGIFQ-UHFFFAOYSA-N |
| Synonym | 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid |
| PubChem CID | 1369 |
| ChEBI | CHEBI:39708 |
| IUPAC Name | 8-anilinonaphthalene-1-sulfonic acid |
| SMILES | C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O |
Chemical Identifiers
| 82-76-8 | |
| 299.344 | |
| FWEOQOXTVHGIFQ-UHFFFAOYSA-N | |
| 1369 | |
| 8-anilinonaphthalene-1-sulfonic acid |
| C16H13NO3S | |
| MFCD00003998 | |
| 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid | |
| CHEBI:39708 | |
| C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O |
Specifications
| 82-76-8 | |
| Black-Gray | |
| MFCD00003998 | |
| 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid | |
| C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O | |
| 299.344 | |
| CHEBI:39708 | |
| ≥95.0% (HPLC,T) | |
| 8-Anilino-1-naphthalenesulfonic Acid |
| 217°C | |
| C16H13NO3S | |
| 25 g | |
| FWEOQOXTVHGIFQ-UHFFFAOYSA-N | |
| 8-anilinonaphthalene-1-sulfonic acid | |
| 1369 | |
| 299.34 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0535
TSCA : Yes