missing translation for 'onlineSavingsMsg'
Learn More

8-Anilino-1-naphthalenesulfonic Acid 95.0+%, TCI America™

Catalog No. A047225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A047225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A047225G Supplier TCI America Supplier No. A047225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82-76-8
Molecular Formula C16H13NO3S
Molecular Weight (g/mol) 299.344
MDL Number MFCD00003998
InChI Key FWEOQOXTVHGIFQ-UHFFFAOYSA-N
Synonym 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid
PubChem CID 1369
ChEBI CHEBI:39708
IUPAC Name 8-anilinonaphthalene-1-sulfonic acid
SMILES C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O

Specifications

Melting Point 217°C
Color Black-Gray
Quantity 25 g
Formula Weight 299.34
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Anilino-1-naphthalenesulfonic Acid
EINECSNumber (4)-0535
TSCA Yes