Learn More
8-Aminoquinoline, 98+%
CAS: 578-66-5 | C9H8N2 | 144.18 g/mol
$125.57 - $194.50
Chemical Identifiers
| CAS | 578-66-5 |
|---|---|
| Molecular Formula | C9H8N2 |
| Molecular Weight (g/mol) | 144.18 |
| MDL Number | MFCD00006809 |
| InChI Key | WREVVZMUNPAPOV-UHFFFAOYSA-N |
| Synonym | 8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline |
| PubChem CID | 11359 |
| IUPAC Name | quinolin-8-amine |
| SMILES | C1=CC2=C(C(=C1)N)N=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC123710050
|
Thermo Scientific Chemicals
123710050 |
5 g |
Each for $125.57
|
|
|||||
|
AC123710100
|
Thermo Scientific Chemicals
123710100 |
10 g |
Each for $194.50
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 578-66-5 | |
| 144.18 | |
| WREVVZMUNPAPOV-UHFFFAOYSA-N | |
| 11359 | |
| C1=CC2=C(C(=C1)N)N=CC=C2 |
| C9H8N2 | |
| MFCD00006809 | |
| 8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline | |
| quinolin-8-amine |
Specifications
| 578-66-5 | |
| 100.0 | |
| Beige-Brown to Green | |
| Authentic | |
| Glass bottle | |
| MFCD00006809 | |
| 22, 450 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in ethanol, insoluble in ether | |
| C1=CC2=C(C(=C1)N)N=CC=C2 | |
| 144.18 | |
| 144.18 | |
| Crystalline Powder |
| 98.0 | |
| 62.0°C to 65.0°C | |
| 174.0°C (26.0 mmHg) | |
| 98% min. (GC) | |
| C9H8N2 | |
| 5 g | |
| 8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline | |
| WREVVZMUNPAPOV-UHFFFAOYSA-N | |
| quinolin-8-amine | |
| 11359 | |
| 98+% | |
| 8-Aminoquinoline |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Suspected of causing genetic defects.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause damage to organs.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 209-427-9
RUO – Research Use Only