missing translation for 'onlineSavingsMsg'
Learn More

8-Aminoquinoline, 98+%

Catalog No. AAA1675203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1675203 1 g
AAA1675206 5 g
AAA1675214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1675203 Supplier Thermo Scientific Chemicals Supplier No. A1675203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 578-66-5 | C9H8N2 | 144.177 g/mol

8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Aminoquinoline has been used in the preparation of base-stabilized terminal borylene complex of osmium. It is also used in the spectrophotometric determination of bivalent palladium.

Solubility
Soluble in water.

Notes
Air Sensitive. Store away from air under dry inert gas. Incompatible with air and oxidizing agents.

Chemical Identifiers

CAS 578-66-5
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006809
InChI Key WREVVZMUNPAPOV-UHFFFAOYSA-N
Synonym 8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
PubChem CID 11359
IUPAC Name quinolin-8-amine
SMILES C1=CC2=C(C(=C1)N)N=CC=C2

Specifications

Melting Point 63°C to 67°C
Boiling Point 174°C (26 mmHg)
Quantity 1 g
Beilstein 114474
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 144.18
Percent Purity ≥98%
Chemical Name or Material 8-Aminoquinoline
Hazard Category H302+H312+H332-H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 209-427-9
RTECSNumber VA9627000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only