missing translation for 'onlineSavingsMsg'
Learn More

8-Aminooctanoic acid, 99%

Catalog No. p-7023614
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1735303 1 g
AAA1735306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1735303 Supplier Thermo Scientific Chemicals Supplier No. A1735303
Only null left

CAS: 1002-57-9 | C8H17NO2 | 159.23 g/mol

8-Aminooctanoic acid is involved in the preparation of lactams using enzyme catalyzed aminolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Aminooctanoic acid is involved in the preparation of lactams using enzyme catalyzed aminolysis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.

Chemical Identifiers

CAS 1002-57-9
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00008245
InChI Key UQXNEWQGGVUVQA-UHFFFAOYSA-N
Synonym 8-aminocaprylic acid, octanoic acid, 8-amino, omega-aminocaprylic acid, 8-amino caprylic acid, 8-amino-octanoic acid, .omega.-aminocaprylic acid, 8-amino capylic acid, 8ac, 8-aminocapylic acid, pubchem13165
PubChem CID 66085
IUPAC Name 8-aminooctanoic acid
SMILES NCCCCCCCC(O)=O

Specifications

Melting Point 193°C to 196°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 159.23
Percent Purity 99%
Chemical Name or Material 8-Aminooctanoic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-687-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only