accessibility menu, dialog, popup

UserName

CAS RN 1002-57-9

IUPAC Name: 8-aminooctanoic acid
Synonyms: 8-aminooctanoic acid
Molecular Weight (g/mol): 159.23
Molecular Formula: C8H17NO2
InChi Key: UQXNEWQGGVUVQA-UHFFFAOYSA-N
SMILES: NCCCCCCCC(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)