missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-2-naphthol, 98%

Catalog No. p-7024549
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1940403 1 g
AAA1940406 5 g
AAA1940414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1940403 Supplier Thermo Scientific Chemicals Supplier No. A1940403
Only null left

CAS: 118-46-7 | C10H9NO | 159.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-46-7
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00004031
InChI Key KVHHMYZBFBSVDI-UHFFFAOYSA-N
Synonym 1-amino-7-naphthol, 8-amino-2-naphthol, 2-naphthalenol, 8-amino, 1-amino-7-hydroxynaphthalene, 7-hydroxy-1-naphthylamine, 2-naphthol, 8-amino, 8-amino-2-naphtol, 8-aminonaphth-2-ol, 1-amino-7-naphthol;, 8-amino-2-naphthaleno
PubChem CID 8358
IUPAC Name 8-aminonaphthalen-2-ol
SMILES NC1=C2C=C(O)C=CC2=CC=C1

Specifications

Melting Point 206°C to 210°C
Quantity 1 g
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 8-Amino-2-naphthol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-252-4
RTECSNumber QL3331000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only