missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt monohydrate, 80%, tech., Thermo Scientific™

$45.63 - $1,015.85

Chemical Identifiers

CAS 5460-09-3
Molecular Formula C10H8NNaO7S2·H2O
Molecular Weight (g/mol) 359.32
MDL Number MFCD00150460
InChI Key QPILZZVXGUNELN-UHFFFAOYSA-M
Synonym h acid sodium, 1-amino-8-naphthol-3,6-disulfonic acid monosodiumsalt, 4-amino-5-hydroxynaphthalene-2,7-disulfonic acid 2-sodium salt
PubChem CID 102116466
IUPAC Name sodium;8-amino-3,6-disulfonaphthalen-1-olate
SMILES C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)N)[O-])S(=O)(=O)O.[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC400840050
View Documents Promotion Details
N/A 5g Glass bottle
Each for $45.63
Only null left
 
AC400841000
View Documents Promotion Details
N/A 100g Plastic bottle
Each for $188.28
Only null left
 
AC400845000
View Documents Promotion Details
N/A 500g Plastic Bottle
Each for $1,015.85
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5460-09-3
359.32
QPILZZVXGUNELN-UHFFFAOYSA-M
102116466
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)N)[O-])S(=O)(=O)O.[Na+]
C10H8NNaO7S2·H2O
MFCD00150460
h acid sodium, 1-amino-8-naphthol-3,6-disulfonic acid monosodiumsalt, 4-amino-5-hydroxynaphthalene-2,7-disulfonic acid 2-sodium salt
sodium;8-amino-3,6-disulfonaphthalen-1-olate

Specifications

5460-09-3
100.0
Authentic
C10H8NNaO7S2·H2O
h acid sodium, 1-amino-8-naphthol-3,6-disulfonic acid monosodiumsalt, 4-amino-5-hydroxynaphthalene-2,7-disulfonic acid 2-sodium salt
QPILZZVXGUNELN-UHFFFAOYSA-M
sodium;8-amino-3,6-disulfonaphthalen-1-olate
102116466
80%
8-Amino-1-naphthol-3, 6-disulfonic acid, monosodium salt monohydrate
75.0
4 % to 6 % (1 g, 150°C, 4 hrs)
75% min. (HPLC)
MFCD00150460
Solubility in water: slightly soluble.
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)N)[O-])S(=O)(=O)O.[Na+]
359.32
359.32
Technical
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-736-4

SDS
Documents

Documents

RUO – Research Use Only