missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt monohydrate, 80%, tech., Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 400841000

Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Catalog No. AC400841000


Only null left

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5460-09-3
359.32
QPILZZVXGUNELN-UHFFFAOYSA-M
62580
[Na+].NC1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S([O-])(=O)=O
C10H8NNaO7S2·H2O
MFCD00150460
piperonyl isobutyrate, unii-y8hbr1acya, y8hbr1acya, piperonyl 2-methylpropanoate, heliotropyl 2-methylpropanoate, isobutyric acid piperonyl ester, propanoic acid, 2-methyl-, 1,3-benzodioxol-5-ylmethyl ester, 1,3-benzodioxol-5-ylmethyl isobutyrate, 3,4-methylenedioxybenzyl 2-methylpropanoate, isobutyric acid 3,4-methylenedioxybenzyl ester
1,3-benzodioxol-5-ylmethyl 2-methylpropanoate

Specifications

8-Amino-1-naphthol-3, 6-disulfonic acid, monosodium salt monohydrate
75.0
4 % to 6 % (1 g, 150°C, 4 hrs)
75% min. (HPLC)
C10H8NNaO7S2·H2O
MFCD00150460
Solubility in water: slightly soluble.
[Na+].NC1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S([O-])(=O)=O
359.32
359.32
Technical
5460-09-3
100.0
Authentic
Plastic bottle
100g
piperonyl isobutyrate, unii-y8hbr1acya, y8hbr1acya, piperonyl 2-methylpropanoate, heliotropyl 2-methylpropanoate, isobutyric acid piperonyl ester, propanoic acid, 2-methyl-, 1,3-benzodioxol-5-ylmethyl ester, 1,3-benzodioxol-5-ylmethyl isobutyrate, 3,4-methylenedioxybenzyl 2-methylpropanoate, isobutyric acid 3,4-methylenedioxybenzyl ester
QPILZZVXGUNELN-UHFFFAOYSA-M
1,3-benzodioxol-5-ylmethyl 2-methylpropanoate
62580
80%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-736-4

SDS
Documents

Documents

Promotions

Promotions