missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic Acid 97.0+%, TCI America™
Supplier: TCI America F07191G
Specifications
| 8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic Acid | |
| White-Yellow | |
| MFCD01321015 | |
| 2-2-fmoc-amino ethoxy ethoxy acetic acid, fmoc-nh-peg2-ch2cooh, fmoc-aeea, 1-9h-fluoren-9-yl-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, 8-fmoc-amino-3,6-dioxaoctanoic acid, fmoc-adoa, fmoc-aeeac-oh, fmoc-amino-3,6 dioxaoctanoic acid, 8-fmoc-amino-3,6-dioxa-n-octanoic acid | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCOCCOCC(=O)O | |
| 385.416 | |
| 385.42 | |
| Crystalline Powder |
| 166108-71-0 | |
| C21H23NO6 | |
| 1 g | |
| XQPYRJIMPDBGRW-UHFFFAOYSA-N | |
| 2-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]acetic acid | |
| 2756092 | |
| ≥97.0% (HPLC) |
Chemical Identifiers
| 166108-71-0 | |
| 385.416 | |
| XQPYRJIMPDBGRW-UHFFFAOYSA-N | |
| 2756092 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCOCCOCC(=O)O |
| C21H23NO6 | |
| MFCD01321015 | |
| 2-2-fmoc-amino ethoxy ethoxy acetic acid, fmoc-nh-peg2-ch2cooh, fmoc-aeea, 1-9h-fluoren-9-yl-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, 8-fmoc-amino-3,6-dioxaoctanoic acid, fmoc-adoa, fmoc-aeeac-oh, fmoc-amino-3,6 dioxaoctanoic acid, 8-fmoc-amino-3,6-dioxa-n-octanoic acid | |
| 2-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]acetic acid |
Safety and Handling
TSCA : No