missing translation for 'onlineSavingsMsg'
Learn More

8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic Acid 97.0+%, TCI America™

Catalog No. F0719200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0719200MG 200 mg
F07191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0719200MG Supplier TCI America Supplier No. F0719200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 166108-71-0
Molecular Formula C21H23NO6
Molecular Weight (g/mol) 385.416
MDL Number MFCD01321015
InChI Key XQPYRJIMPDBGRW-UHFFFAOYSA-N
Synonym 2-2-fmoc-amino ethoxy ethoxy acetic acid, fmoc-nh-peg2-ch2cooh, fmoc-aeea, 1-9h-fluoren-9-yl-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, 8-fmoc-amino-3,6-dioxaoctanoic acid, fmoc-adoa, fmoc-aeeac-oh, fmoc-amino-3,6 dioxaoctanoic acid, 8-fmoc-amino-3,6-dioxa-n-octanoic acid
PubChem CID 2756092
IUPAC Name 2-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]acetic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCOCCOCC(=O)O

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 385.42
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic Acid