missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ 7X™ Laboratory Detergents
Safe, effective cleaning solutions for glass and plastic labware
Supplier: MP Biomedicals™ 097667095
Description
7X cleaning solution was developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
- Effective and safe
- Completely drains from glassware in seconds
- Eliminates residue leaving no interfering fluorescence
- pH neutral, and no need for pH adjustment at any concentration
- No etch to glass or plastic labware at any concentration
Specifications
| Liquid | |
| 7X | |
| Use on Glass, Plastic, benchtop equipment and tubing |
| 6 to 7.5 | |
| 15°C to 30°C | |
| 5 gal. |