missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ 7712 Series PVC Coated Gloves
PVC coated, multi-purpose gloves with soft cotton liner are designed for all industrial applications and protect against a range of hazards including chemicals, oils and abrasion.
Supplier: SHOWA™ 7712R10
Description
Both styles are ideal for working with chemicals, oil, or grease in refining, mining, automotive subassembly, or construction
- Fully coated PVC with smooth or rough finish
- Cotton jersey lined Sanitized™
- Recyclable
- Washable
Specifications
| CFIA | |
| Yes | |
| Large | |
| Gauntlet | |
| Cotton | |
| 7712R-10 | |
| 12 in. |
| Black | |
| PVC | |
| Chemical spray and treatment, coating preparation, laboratory, pharma and analysis, washing and cleaning | |
| Rough | |
| Lined | |
| 7712R Series | |
| 30.5 cm |