missing translation for 'onlineSavingsMsg'
Learn More

7-(Trifluoromethyl)quinoline, 97%

Catalog No. p-7030216
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27771MD 250 mg
AAH2777103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27771MD Supplier Thermo Scientific Chemicals Supplier No. H27771MD

CAS: 325-14-4 | C10H6F3N | 197.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 325-14-4
Molecular Formula C10H6F3N
Molecular Weight (g/mol) 197.16
MDL Number MFCD00833760
InChI Key CMMSEFHVUYEEDY-UHFFFAOYSA-N
Synonym 7-trifluoromethyl quinoline, 7-cf3 quinoline, 7-trrfluoromethylquinoline, pubchem7218, 7-trifluoromethyl-quinoline, acmc-20a40f, quinoline,7-trifluoromethyl, quinoline, 7-trifluoromethyl
PubChem CID 459614
IUPAC Name 7-(trifluoromethyl)quinoline
SMILES C1=CC2=C(C=C(C=C2)C(F)(F)F)N=C1

Specifications

Melting Point 65°C to 66°C
Quantity 250 mg
Formula Weight 197.16
Percent Purity 97%
Chemical Name or Material 7-(Trifluoromethyl)quinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only