accessibility menu, dialog, popup

UserName

CAS RN 325-14-4

IUPAC Name: 7-(trifluoromethyl)quinoline
Synonyms: 7-CF3 quinoline
Molecular Weight (g/mol): 197.16
Molecular Formula: C10H6F3N
InChi Key: CMMSEFHVUYEEDY-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC2=NC=CC=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)