missing translation for 'onlineSavingsMsg'
Learn More

7-(Trifluoromethyl)isatin 98.0+%, TCI America™

Catalog No. T24571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24571G 1 g
T24575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24571G Supplier TCI America Supplier No. T24571G

Chemical Identifiers

CAS 391-12-8
Molecular Formula C9H4F3NO2
Molecular Weight (g/mol) 215.131
MDL Number MFCD01248408
InChI Key MXLDJTXXAYVWDF-UHFFFAOYSA-N
Synonym 7-(Trifluoromethyl)indole-2,3-dione, 7-(Trifluoromethyl)-2,3-indolinedione
PubChem CID 604575
IUPAC Name 7-(trifluoromethyl)-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)C(F)(F)F)NC(=O)C2=O

Specifications

Melting Point 193°C
Quantity 1 g
Formula Weight 215.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Trifluoromethyl)isatin