accessibility menu, dialog, popup

UserName

CAS RN 391-12-8

FFFOOHN

CAS RN 391-12-8

IUPAC Name: 7-(trifluoromethyl)-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 7-(trifluoromethyl)-1H-indole-2,3-dione
Molecular Weight (g/mol): 215.13
Molecular Formula: C9H4F3NO2
InChi Key: MXLDJTXXAYVWDF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC2=C1NC(=O)C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results