missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Tetradecyne 97.0+%, TCI America™
Supplier: TCI America T07625ML
Specifications
| 7-Tetradecyne | |
| Yellow | |
| C14H26 | |
| 5 mL | |
| CCCCCCC#CCCCCCC | |
| 194.362 | |
| 194.36 | |
| Liquid |
| 35216-11-6 | |
| 144°C | |
| MFCD00041662 | |
| AFNWSIIBAYUTTL-UHFFFAOYSA-N | |
| tetradec-7-yne | |
| 141979 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 35216-11-6 | |
| 194.362 | |
| AFNWSIIBAYUTTL-UHFFFAOYSA-N | |
| tetradec-7-yne |
| C14H26 | |
| MFCD00041662 | |
| 141979 | |
| CCCCCCC#CCCCCCC |
Safety and Handling
TSCA : No