missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Tetradecyne 97.0+%, TCI America™
$301.98 - $907.79
Chemical Identifiers
| CAS | 35216-11-6 |
|---|---|
| Molecular Formula | C14H26 |
| Molecular Weight (g/mol) | 194.362 |
| MDL Number | MFCD00041662 |
| InChI Key | AFNWSIIBAYUTTL-UHFFFAOYSA-N |
| PubChem CID | 141979 |
| IUPAC Name | tetradec-7-yne |
| SMILES | CCCCCCC#CCCCCCC |
Chemical Identifiers
| 35216-11-6 | |
| 194.362 | |
| AFNWSIIBAYUTTL-UHFFFAOYSA-N | |
| tetradec-7-yne |
| C14H26 | |
| MFCD00041662 | |
| 141979 | |
| CCCCCCC#CCCCCCC |
Specifications
| 35216-11-6 | |
| 144°C | |
| MFCD00041662 | |
| AFNWSIIBAYUTTL-UHFFFAOYSA-N | |
| tetradec-7-yne | |
| 141979 | |
| ≥97.0% (GC) | |
| 7-Tetradecyne |
| Yellow | |
| C14H26 | |
| 5 mL | |
| CCCCCCC#CCCCCCC | |
| 194.362 | |
| 194.36 | |
| Liquid |
Safety and Handling
TSCA : No