missing translation for 'onlineSavingsMsg'
Learn More

7-Tetradecene 97.0+%, TCI America™

Catalog No. T07455ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07455ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T07455ML Supplier TCI America Supplier No. T07455ML
Only null left

Chemical Identifiers

CAS 10374-74-0
Molecular Formula C14H28
Molecular Weight (g/mol) 196.378
MDL Number MFCD00009550
InChI Key UBDIXSAEHLOROW-BUHFOSPRSA-N
PubChem CID 5364651
IUPAC Name (E)-tetradec-7-ene
SMILES CCCCCCC=CCCCCCC

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 196.38
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 7-Tetradecene
EINECSNumber (2)-0027
TSCA No