accessibility menu, dialog, popup

UserName

CAS RN 10374-74-0

CH3CH3(E)

CAS RN 10374-74-0

IUPAC Name: (7E)-tetradec-7-ene
Synonyms: (7E)-tetradec-7-ene
Molecular Weight (g/mol): 196.38
Molecular Formula: C14H28
InChi Key: UBDIXSAEHLOROW-BUHFOSPRSA-N
SMILES: CCCCCC\C=C\CCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results