missing translation for 'onlineSavingsMsg'
Learn More

7-Phenyl-1-heptanol, 97%

Catalog No. AAL1253306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1253306 5 g
AAL1253303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1253306 Supplier Thermo Scientific Chemicals Supplier No. L1253306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3208-25-1 | C13H20O | 192.302 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3208-25-1
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD00037315
InChI Key UXMUSYTXSNVRMW-UHFFFAOYSA-N
Synonym benzeneheptanol, 7-phenyl-1-heptanol, 7-phenylheptanol, phenylheptyl alcohol, 7-phenyl-n-heptanol, acmc-20alwo, 7-phenylheptyl alcohol
PubChem CID 154271
IUPAC Name 7-phenylheptan-1-ol
SMILES C1=CC=C(C=C1)CCCCCCCO

Specifications

Density 0.962
Boiling Point 115°C to 117°C (0.5 mmHg)
Refractive Index 1.508
Quantity 5 g
Beilstein 1865431
Formula Weight 192.3
Percent Purity 97%
Chemical Name or Material 7-Phenyl-1-heptanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only