accessibility menu, dialog, popup

UserName

HO

CAS RN 3208-25-1

IUPAC Name: 7-phenylheptan-1-ol
Synonyms: 7-phenylheptan-1-ol
Molecular Weight (g/mol): 192.3
Molecular Formula: C13H20O
InChi Key: UXMUSYTXSNVRMW-UHFFFAOYSA-N
SMILES: OCCCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)