missing translation for 'onlineSavingsMsg'
Learn More

7-Nitroindole 98.0+%, TCI America™

Catalog No. N06851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06851G 1 g
N06855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06851G Supplier TCI America Supplier No. N06851G
Only null left

Chemical Identifiers

CAS 6960-42-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00005683
InChI Key LZJGQIVWUKFTRD-UHFFFAOYSA-N
Synonym 7-nitroindole, 1h-indole, 7-nitro, indole, 7-nitro, 7-nitro indole, pubchem7441, acmc-1b2jq, 5-20-07-00044 beilstein handbook reference, ksc353k7l, 1h-indole, 7-nitro-9ci
PubChem CID 23396
IUPAC Name 7-nitro-1H-indole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2

Specifications

Melting Point 97°C
Color Yellow
Quantity 1 g
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Nitroindole
RTECSNumber NM1169000
TSCA No