Learn More
7-Nitroindole-2-carboxylic acid, 96%
CAS: 6960-45-8 | C9H6N2O4 | 206.157 g/mol
$195.95 - $732.60
Chemical Identifiers
| CAS | 6960-45-8 |
|---|---|
| Molecular Formula | C9H6N2O4 |
| Molecular Weight (g/mol) | 206.157 |
| MDL Number | MFCD00044720 |
| InChI Key | BIUCOFQROHIAEO-UHFFFAOYSA-N |
| Synonym | 7-nitro-1h-indole-2-carboxylic acid, 7-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 7-nitro, 7-nitroindole-2-carboxylicacid, dna base excision repair pathway inhibitor, maybridge1_004786, indole-2-carboxylic acid, 7-nitro, pubchem9857, acmc-1b8pb, 1h-indole-2-carboxylicacid, 7-nitro |
| PubChem CID | 81409 |
| SMILES | C1=CC2=C(C(=C1)[N+](=O)[O-])NC(=C2)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2078603
|
Thermo Scientific Chemicals
B2078603 |
1 g |
Each for $195.95
|
|
|||||
|
AAB2078606
|
Thermo Scientific Chemicals
B2078606 |
5 g |
Each for $732.60
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6960-45-8 | |
| 206.157 | |
| BIUCOFQROHIAEO-UHFFFAOYSA-N | |
| 81409 |
| C9H6N2O4 | |
| MFCD00044720 | |
| 7-nitro-1h-indole-2-carboxylic acid, 7-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 7-nitro, 7-nitroindole-2-carboxylicacid, dna base excision repair pathway inhibitor, maybridge1_004786, indole-2-carboxylic acid, 7-nitro, pubchem9857, acmc-1b8pb, 1h-indole-2-carboxylicacid, 7-nitro | |
| C1=CC2=C(C(=C1)[N+](=O)[O-])NC(=C2)C(=O)O |
Specifications
| 6960-45-8 | |
| C9H6N2O4 | |
| 1 g | |
| BIUCOFQROHIAEO-UHFFFAOYSA-N | |
| 206.157 | |
| 206.16 | |
| 7-Nitroindole-2-carboxylic acid |
| 264°C to 266°C | |
| MFCD00044720 | |
| 7-nitro-1h-indole-2-carboxylic acid, 7-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 7-nitro, 7-nitroindole-2-carboxylicacid, dna base excision repair pathway inhibitor, maybridge1_004786, indole-2-carboxylic acid, 7-nitro, pubchem9857, acmc-1b8pb, 1h-indole-2-carboxylicacid, 7-nitro | |
| C1=CC2=C(C(=C1)[N+](=O)[O-])NC(=C2)C(=O)O | |
| 81409 | |
| 96% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335-H500
EINECSNumber : 230-154-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only