missing translation for 'onlineSavingsMsg'
Learn More

7-Nitroindole-2-carboxylic acid, 96%

Catalog No. AAB2078606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2078606 5 g
AAB2078603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2078606 Supplier Thermo Scientific Chemicals Supplier No. B2078606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6960-45-8 | C9H6N2O4 | 206.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6960-45-8
Molecular Formula C9H6N2O4
Molecular Weight (g/mol) 206.157
MDL Number MFCD00044720
InChI Key BIUCOFQROHIAEO-UHFFFAOYSA-N
Synonym 7-nitro-1h-indole-2-carboxylic acid, 7-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 7-nitro, 7-nitroindole-2-carboxylicacid, dna base excision repair pathway inhibitor, maybridge1_004786, indole-2-carboxylic acid, 7-nitro, pubchem9857, acmc-1b8pb, 1h-indole-2-carboxylicacid, 7-nitro
PubChem CID 81409
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NC(=C2)C(=O)O

Specifications

Melting Point 264°C to 266°C
Quantity 5 g
Formula Weight 206.16
Percent Purity 96%
Chemical Name or Material 7-Nitroindole-2-carboxylic acid
Hazard Category H302+H312-H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 230-154-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only