missing translation for 'onlineSavingsMsg'
Learn More

7-Methylquinoline, 97%

Catalog No. AA4247501
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4247501 0.25 g
AA4247503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4247501 Supplier Thermo Scientific Chemicals Supplier No. 04247501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 612-60-2 | C10H9N | 143.189 g/mol

7-methylquinoline has been bioassayed as tumor initiator on the skin of Sencar mice.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 612-60-2
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.189
MDL Number MFCD00006805
InChI Key KDYVCOSVYOSHOL-UHFFFAOYSA-N
Synonym quinoline, 7-methyl, m-toluquinoline, unii-eow995k43c, 7-methyl quinoline, ccris 2895, quinolinol,7-methyl, 7-methyl-quinoline, 7-methylchinolin, 7-methylchinolin;, pubchem5811
PubChem CID 11927
IUPAC Name 7-methylquinoline
SMILES CC1=CC2=C(C=CC=N2)C=C1

Specifications

Density 1.07 g/mL
Boiling Point 251°C to 253°C
Flash Point >110°C (230°F)
Quantity 0.25 g
Beilstein 110317
Solubility Information Insoluble in water.
Formula Weight 143.19
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 7-Methylquinoline
Hazard Category H302-H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 210-316-2
RTECSNumber VC0560000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only