missing translation for 'onlineSavingsMsg'
Learn More

7-Methylbenz[a]anthracene 97.0+%, TCI America™

Catalog No. M0248100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0248100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0248100MG Supplier TCI America Supplier No. M0248100MG
Only null left

Chemical Identifiers

CAS 2541-69-7
Molecular Formula C19H14
Molecular Weight (g/mol) 242.321
MDL Number MFCD00059500
InChI Key DIIFUCUPDHMNIV-UHFFFAOYSA-N
PubChem CID 17347
IUPAC Name 7-methylbenzo[a]anthracene
SMILES CC1=C2C=CC3=CC=CC=C3C2=CC4=CC=CC=C14

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 100 mg
Formula Weight 242.32
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methylbenz[a]anthracene
RTECSNumber CX1575000
TSCA No