missing translation for 'onlineSavingsMsg'
Learn More

7-Methyl lumazine

Catalog No. AAJ64389MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64389MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64389MB Supplier Thermo Scientific Chemicals Supplier No. J64389MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13401-38-2 | C7H6N4O2 | 178.15 g/mol

A methyl-substituted imidazo[1,2-a]pyrazine derivative

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A methyl-substituted imidazo[1,2-a]pyrazine derivative with anti-inflammatory, analgesic and ulcerogenic activities.

Chemical Identifiers

CAS 13401-38-2
Molecular Formula C7H6N4O2
Molecular Weight (g/mol) 178.15
MDL Number MFCD19688352
InChI Key WIXGFZWEUAUYGB-UHFFFAOYSA-N
Synonym 7-methyl lumazine, 7-methyllumazine, 7-methyllumizine, lumazine, 7-methyl, 7-methylpteridine-2,4 1h,3h-dione, 7-methylpteridine-2,4-diol, 2,4 1h,3h-pteridinedione, 7-methyl, 7-methyl-1,3-dihydropteridine-2,4-dione, 7-methyl-pteridine-2,4-diol, methyllumazin isomerengemisch
PubChem CID 5374589
IUPAC Name 7-methyl-1H-pteridine-2,4-dione
SMILES CC1=CN=C2C(=O)NC(=O)NC2=N1

Specifications

Quantity 25 mg
Formula Weight 178.15
Physical Form Powder
Chemical Name or Material 7-Methyl lumazine
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only