accessibility menu, dialog, popup

UserName

CAS RN 13401-38-2

IUPAC Name: 7-methyl-1,2,3,4-tetrahydropteridine-2,4-dione
Synonyms: 7-methyl-1,3-dihydropteridine-2,4-dione
Molecular Weight (g/mol): 178.15
Molecular Formula: C7H6N4O2
InChi Key: WIXGFZWEUAUYGB-UHFFFAOYSA-N
SMILES: CC1=CN=C2C(=O)NC(=O)NC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results