missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene 95.0+%, TCI America™
$137.94 - $452.97
Chemical Identifiers
| CAS | 84030-20-6 |
|---|---|
| Molecular Formula | C8H15N3 |
| Molecular Weight (g/mol) | 153.229 |
| MDL Number | MFCD00043004 |
| InChI Key | OEBXWWBYZJNKRK-UHFFFAOYSA-N |
| Synonym | 1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine, MTBD |
| PubChem CID | 123583 |
| IUPAC Name | 1-methyl-2,3,4,6,7,8-hexahydropyrimido[1,2-a]pyrimidine |
| SMILES | CN1CCCN2C1=NCCC2 |
Chemical Identifiers
| 84030-20-6 | |
| 153.229 | |
| OEBXWWBYZJNKRK-UHFFFAOYSA-N | |
| 123583 | |
| CN1CCCN2C1=NCCC2 |
| C8H15N3 | |
| MFCD00043004 | |
| 1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine, MTBD | |
| 1-methyl-2,3,4,6,7,8-hexahydropyrimido[1,2-a]pyrimidine |
Specifications
| 84030-20-6 | |
| C8H15N3 | |
| 1 g | |
| 1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine, MTBD | |
| CN1CCCN2C1=NCCC2 | |
| 153.229 | |
| 153.23 | |
| Liquid |
| 79°C | |
| MFCD00043004 | |
| 3267 | |
| OEBXWWBYZJNKRK-UHFFFAOYSA-N | |
| 1-methyl-2,3,4,6,7,8-hexahydropyrimido[1,2-a]pyrimidine | |
| 123583 | |
| ≥95.0% (GC,T) | |
| 7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator