accessibility menu, dialog, popup

UserName

CAS RN 84030-20-6

CH3NNN

CAS RN 84030-20-6

IUPAC Name: 1-methyl-1H,2H,3H,4H,6H,7H,8H-[1,3]diazino[1,2-a]pyrimidine
Synonyms: 1-methyl-2H,3H,4H,6H,7H,8H-[1,3]diazino[1,2-a]pyrimidine
Molecular Weight (g/mol): 153.23
Molecular Formula: C8H15N3
InChi Key: OEBXWWBYZJNKRK-UHFFFAOYSA-N
SMILES: CN1CCCN2CCCN=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results