missing translation for 'onlineSavingsMsg'
Learn More

7-Methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 95%, Thermo Scientific Chemicals

Catalog No. p-7032633
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3416803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3416803 Supplier Thermo Scientific Chemicals Supplier No. H3416803

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 274676-47-0
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD06739289
InChI Key HBWCJMBLUCNJIS-UHFFFAOYSA-N
Synonym 2-methyl-5,6,7,8-tetrahydro-1,8-naphthyridine, 7-methyl-1,2,3,4-tetrahydro-1,8 naphthyridine, 7-methyl-1,2,3,4-tetrahydropyridino 2,3-b pyridine, 1,2,3,4-tetrahydro-7-methyl-1,8-naphthyridine, 1,8-naphthyridine, 1,2,3,4-tetrahydro-7-methyl-9ci
PubChem CID 22319466
IUPAC Name 7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine
SMILES CC1=NC2=C(CCCN2)C=C1

Specifications

Melting Point 88°C to 91°C
Quantity 1 g
Formula Weight 148.21
Percent Purity 95%
Chemical Name or Material 7-Methyl-1,2,3,4-tetrahydro-1,8-naphthyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only