CAS RN 274676-47-0
IUPAC Name:
7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine
Synonyms:
2-methyl-5,6,7,8-tetrahydro-1,8-naphthyridine
Molecular Weight (g/mol):
148.21
Molecular Formula:
C9H12N2
InChi Key:
HBWCJMBLUCNJIS-UHFFFAOYSA-N
SMILES:
CC1=NC2=C(CCCN2)C=C1
1
–
1
of
1
results
1
7-Methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 95%, Thermo Scientific Chemicals
CAS: 274676-47-0 Molecular Formula: C9H12N2 Molecular Weight (g/mol): 148.209 MDL Number: MFCD06739289 InChI Key: HBWCJMBLUCNJIS-UHFFFAOYSA-N Synonym: 2-methyl-5,6,7,8-tetrahydro-1,8-naphthyridine,7-methyl-1,2,3,4-tetrahydro-1,8 naphthyridine,7-methyl-1,2,3,4-tetrahydropyridino 2,3-b pyridine,1,2,3,4-tetrahydro-7-methyl-1,8-naphthyridine,1,8-naphthyridine, 1,2,3,4-tetrahydro-7-methyl-9ci PubChem CID: 22319466 IUPAC Name: 7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine SMILES: CC1=NC2=C(CCCN2)C=C1