missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxycoumarin-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7133103
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2233100MG 100 mg
M22331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2233100MG Supplier TCI America Supplier No. M2233100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20300-59-8
Molecular Formula C11H8O5
Molecular Weight (g/mol) 220.18
MDL Number MFCD00452770
InChI Key VEEGNDSSWAOLFN-UHFFFAOYSA-N
PubChem CID 583941
IUPAC Name 7-methoxy-2-oxo-2H-chromene-3-carboxylic acid
SMILES COC1=CC=C2C=C(C(O)=O)C(=O)OC2=C1

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 100 mg
Formula Weight 220.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxycoumarin-3-carboxylic Acid
TSCA No
Recommended Storage Freezer