accessibility menu, dialog, popup

UserName

CAS RN 20300-59-8

IUPAC Name: 7-methoxy-2-oxo-2H-chromene-3-carboxylic acid
Synonyms: 7-methoxy-2-oxochromene-3-carboxylic acid
Molecular Weight (g/mol): 220.18
Molecular Formula: C11H8O5
InChi Key: VEEGNDSSWAOLFN-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(C(O)=O)C(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results