missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Methoxy-4-quinolinol 97.0+%, TCI America™
$169.63 - $665.46
Chemical Identifiers
| CAS | 82121-05-9 |
|---|---|
| Molecular Formula | C10H9NO2 |
| Molecular Weight (g/mol) | 175.187 |
| MDL Number | MFCD00169015 |
| InChI Key | NQUPXNZWBGZRQX-UHFFFAOYSA-N |
| Synonym | 4-Hydroxy-7-methoxyquinoline |
| PubChem CID | 2831995 |
| IUPAC Name | 7-methoxy-1H-quinolin-4-one |
| SMILES | COC1=CC2=C(C=C1)C(=O)C=CN2 |
Chemical Identifiers
| 82121-05-9 | |
| 175.187 | |
| NQUPXNZWBGZRQX-UHFFFAOYSA-N | |
| 2831995 | |
| COC1=CC2=C(C=C1)C(=O)C=CN2 |
| C10H9NO2 | |
| MFCD00169015 | |
| 4-Hydroxy-7-methoxyquinoline | |
| 7-methoxy-1H-quinolin-4-one |
Specifications
| 82121-05-9 | |
| C10H9NO2 | |
| 1 g | |
| NQUPXNZWBGZRQX-UHFFFAOYSA-N | |
| 7-methoxy-1H-quinolin-4-one | |
| 2831995 | |
| ≥97.0% (T) | |
| 7-Methoxy-4-quinolinol |
| 285°C | |
| MFCD00169015 | |
| 4-Hydroxy-7-methoxyquinoline | |
| COC1=CC2=C(C=C1)C(=O)C=CN2 | |
| 175.187 | |
| 175.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No