accessibility menu, dialog, popup

UserName

CH3OONH

CAS RN 82121-05-9

IUPAC Name: 7-methoxy-1,4-dihydroquinolin-4-one
Synonyms: 7-methoxy-1H-quinolin-4-one
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: NQUPXNZWBGZRQX-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(=O)C=CNC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results