missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-4-quinolinol 97.0+%, TCI America™

Catalog No. M19665G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19665G 5 g
M19661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19665G Supplier TCI America Supplier No. M19665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82121-05-9
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00169015
InChI Key NQUPXNZWBGZRQX-UHFFFAOYSA-N
Synonym 4-Hydroxy-7-methoxyquinoline
PubChem CID 2831995
IUPAC Name 7-methoxy-1H-quinolin-4-one
SMILES COC1=CC2=C(C=C1)C(=O)C=CN2

Specifications

Melting Point 285°C
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxy-4-quinolinol