missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-4-methylcoumarin 98.0+%, TCI America™

Catalog No. M13935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13935G Supplier TCI America Supplier No. M13935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2555-28-4
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00009773
InChI Key UDFPKNSWSYBIHO-UHFFFAOYSA-N
Synonym 7-methoxy-4-methylcoumarin, 7-methoxy-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-methoxy-4-methyl, 4-methylherniarin, hymecromone methyl ether, herniarin, 4-methyl, 7-methoxy-4-methyl-chromen-2-one, 4-methyl-7-methoxycoumarin, coumarin,4-methyl-7-methoxy, coumarin, 7-methoxy-4-methyl
PubChem CID 390807
IUPAC Name 7-methoxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxy-4-methylcoumarin