missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-2-tetralone, 95%, Thermo Scientific™

$293.16 - $293.16

Chemical Identifiers

CAS 4133-34-0
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00001730
InChI Key XEAPZXNZOJGVCZ-UHFFFAOYSA-N
Synonym 7-methoxy-2-tetralone, 7-methoxy-3,4-dihydronaphthalen-2 1h-one, 3,4-dihydro-7-methoxy-2 1h-naphthalenone, 7-methoxy-3,4-dihydronaphthalen-2-one, 7-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 7-methoxyl-2-tetralone, 7-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxy-1,3,4-trihydronaphthalen-2-one, 3,4-dihydro-7-methoxynaphthalen-2 1h-one
PubChem CID 77785
IUPAC Name 7-methoxy-3,4-dihydro-1H-naphthalen-2-one
SMILES COC1=CC2=C(CCC(=O)C2)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC294970050
SDS View Documents
Thermo Scientific Chemicals
294970050
5 g Glass bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4133-34-0
176.22
XEAPZXNZOJGVCZ-UHFFFAOYSA-N
77785
COC1=CC2=C(CCC(=O)C2)C=C1
C11H12O2
MFCD00001730
7-methoxy-2-tetralone, 7-methoxy-3,4-dihydronaphthalen-2 1h-one, 3,4-dihydro-7-methoxy-2 1h-naphthalenone, 7-methoxy-3,4-dihydronaphthalen-2-one, 7-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 7-methoxyl-2-tetralone, 7-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxy-1,3,4-trihydronaphthalen-2-one, 3,4-dihydro-7-methoxynaphthalen-2 1h-one
7-methoxy-3,4-dihydro-1H-naphthalen-2-one

Specifications

4133-34-0
100.0
1.1300g/mL
>110°C
94% min. (GC)
C11H12O2
MFCD00001730
1.13
XEAPZXNZOJGVCZ-UHFFFAOYSA-N
7-methoxy-3,4-dihydro-1H-naphthalen-2-one
77785
95%
7-Methoxy-2-tetralone
94.0
Yellow to Orange
124°C to 126°C (1.5 mmHg)
Authentic
Glass bottle
1.5590 to 1.563
5 g
7-methoxy-2-tetralone, 7-methoxy-3,4-dihydronaphthalen-2 1h-one, 3,4-dihydro-7-methoxy-2 1h-naphthalenone, 7-methoxy-3,4-dihydronaphthalen-2-one, 7-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 7-methoxyl-2-tetralone, 7-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxy-1,3,4-trihydronaphthalen-2-one, 3,4-dihydro-7-methoxynaphthalen-2 1h-one
COC1=CC2=C(CCC(=O)C2)C=C1
176.22
176.22
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-954-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only