missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 95%, Thermo Scientific Chemicals

Catalog No. p-7056942
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66188MD 250 mg
AAH6618803 1 g
AAH6618806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66188MD Supplier Thermo Scientific Chemicals Supplier No. H66188MD

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1810-74-8
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
MDL Number MFCD00627122
InChI Key VNIQAUZZZWOJPT-UHFFFAOYSA-N
Synonym 7-methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 2,2,4-trimethyl-7-methoxy-1,2-dihydroquinoline, quinoline, 1,2-dihydro-7-methoxy-2,2,4-trimethyl, 1,2-dihydro-2,2,4-trimethyl-7-methoxyquinoline, 1,2-dihydro-7-methoxy-2,2,4-trimethylquinoline, 7-methoxy-2,2,4-trimethyl-1,2-dihydro-quinoline, quinoline, 7-methoxy-2,2,4-trimethyl-1,2-dihydro
PubChem CID 643478
IUPAC Name 7-methoxy-2,2,4-trimethyl-1H-quinoline
SMILES CC1=CC(NC2=C1C=CC(=C2)OC)(C)C

Specifications

Quantity 250 mg
Beilstein 1426616
Formula Weight 203.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only