CAS RN 1810-74-8
IUPAC Name:
7-methoxy-2,2,4-trimethyl-1,2-dihydroquinoline
Synonyms:
7-methoxy-2,2,4-trimethyl-1H-quinoline
Molecular Weight (g/mol):
203.29
Molecular Formula:
C13H17NO
InChi Key:
VNIQAUZZZWOJPT-UHFFFAOYSA-N
SMILES:
COC1=CC2=C(C=C1)C(C)=CC(C)(C)N2
1
–
1
of
1
results
1
Promotions Available
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 95%, Thermo Scientific Chemicals
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 95%, Thermo Scientific Chemicals
CAS: 1810-74-8 Molecular Formula: C13H17NO Molecular Weight (g/mol): 203.285 MDL Number: MFCD00627122 InChI Key: VNIQAUZZZWOJPT-UHFFFAOYSA-N Synonym: 7-methoxy-2,2,4-trimethyl-1,2-dihydroquinoline,2,2,4-trimethyl-7-methoxy-1,2-dihydroquinoline,quinoline, 1,2-dihydro-7-methoxy-2,2,4-trimethyl,1,2-dihydro-2,2,4-trimethyl-7-methoxyquinoline,1,2-dihydro-7-methoxy-2,2,4-trimethylquinoline,7-methoxy-2,2,4-trimethyl-1,2-dihydro-quinoline,quinoline, 7-methoxy-2,2,4-trimethyl-1,2-dihydro PubChem CID: 643478 IUPAC Name: 7-methoxy-2,2,4-trimethyl-1H-quinoline SMILES: CC1=CC(NC2=C1C=CC(=C2)OC)(C)C