missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-1-tetralone 98.0+%, TCI America™

Catalog No. M20925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20925G 5 g
M209225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20925G Supplier TCI America Supplier No. M20925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6836-19-7
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00001696
InChI Key GABLTKRIYDNDIN-UHFFFAOYSA-N
Synonym 7-methoxy-1-tetralone, 7-methoxyl-1-tetralone, 7-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 7-methoxy-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxytetralin-1-one, 3,4-dihydro-7-methoxy-1 2h-naphthalenone, 7-methoxy-2,3,4-trihydronaphthalen-1-one, 7-methoxytetralone, 7-methoxy-tetralone
PubChem CID 81276
IUPAC Name 7-methoxy-3,4-dihydro-2H-naphthalen-1-one
SMILES COC1=CC2=C(CCCC2=O)C=C1

Specifications

Melting Point 62°C
Color White-Yellow
Quantity 5 g
Formula Weight 176.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxy-1-tetralone