missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Methoxy-1-naphthylacetonitrile 99.0+%, TCI America™
$59.65 - $238.60
Chemical Identifiers
| CAS | 138113-08-3 |
|---|---|
| Molecular Formula | C13H11NO |
| Molecular Weight (g/mol) | 197.24 |
| MDL Number | MFCD08704309 |
| InChI Key | PYJMGUQHJINLLD-UHFFFAOYSA-N |
| Synonym | 7-Methoxy-1-naphthylmethylcyanide |
| PubChem CID | 11264068 |
| IUPAC Name | 2-(7-methoxynaphthalen-1-yl)acetonitrile |
| SMILES | COC1=CC=C2C=CC=C(CC#N)C2=C1 |
Chemical Identifiers
| 138113-08-3 | |
| 197.24 | |
| PYJMGUQHJINLLD-UHFFFAOYSA-N | |
| 11264068 | |
| COC1=CC=C2C=CC=C(CC#N)C2=C1 |
| C13H11NO | |
| MFCD08704309 | |
| 7-Methoxy-1-naphthylmethylcyanide | |
| 2-(7-methoxynaphthalen-1-yl)acetonitrile |
Specifications
| 138113-08-3 | |
| C13H11NO | |
| 1 g | |
| 7-Methoxy-1-naphthylmethylcyanide | |
| COC1=CC=C2C=CC=C(CC#N)C2=C1 | |
| 197.24 | |
| 197.24 | |
| Crystalline Powder |
| 84°C | |
| MFCD08704309 | |
| 3439 | |
| PYJMGUQHJINLLD-UHFFFAOYSA-N | |
| 2-(7-methoxynaphthalen-1-yl)acetonitrile | |
| 11264068 | |
| ≥99.0% (GC) | |
| 7-Methoxy-1-naphthylacetonitrile |
Safety and Handling
TSCA : No