missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-1-naphthylacetonitrile 99.0+%, TCI America™

Catalog No. M24211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24211G 1 g
M24215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24211G Supplier TCI America Supplier No. M24211G

Chemical Identifiers

CAS 138113-08-3
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD08704309
InChI Key PYJMGUQHJINLLD-UHFFFAOYSA-N
Synonym 7-Methoxy-1-naphthylmethylcyanide
PubChem CID 11264068
IUPAC Name 2-(7-methoxynaphthalen-1-yl)acetonitrile
SMILES COC1=CC=C2C=CC=C(CC#N)C2=C1

Specifications

Melting Point 84°C
Quantity 1 g
UN Number 3439
Formula Weight 197.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxy-1-naphthylacetonitrile