missing translation for 'onlineSavingsMsg'
Learn More

7-Isopropyl-1,4-dimethylazulene-3-carboxaldehyde 97.0+%, TCI America™

Catalog No. p-7131659
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0928200MG 200 mg
I09281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0928200MG Supplier TCI America Supplier No. I0928200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3331-47-3
Molecular Formula C16H18O
Molecular Weight (g/mol) 226.32
MDL Number MFCD00275723
InChI Key CDVRGGMPPUFSFH-UHFFFAOYSA-N
Synonym 5-Isopropyl-3,8-dimethylazulene-1-carboxaldehyde
PubChem CID 639851
IUPAC Name 3,8-dimethyl-5-(propan-2-yl)azulene-1-carbaldehyde
SMILES CC(C)C1=CC=C(C)C2=C(C=O)C=C(C)C2=C1

Specifications

Melting Point 83°C
Color Black-Purple
Quantity 200 mg
Formula Weight 226.32
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Isopropyl-1,4-dimethylazulene-3-carboxaldehyde