CAS RN 3331-47-3
IUPAC Name:
3,8-dimethyl-5-(propan-2-yl)azulene-1-carbaldehyde
Synonyms:
5-isopropyl-3,8-dimethylazulene-1-carbaldehyde
Molecular Weight (g/mol):
226.32
Molecular Formula:
C16H18O
InChi Key:
CDVRGGMPPUFSFH-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=C(C)C2=C(C=O)C=C(C)C2=C1
1
–
1
of
1
results
1
7-Isopropyl-1,4-dimethylazulene-3-carboxaldehyde 97.0+%, TCI America™
CAS: 3331-47-3 Molecular Formula: C16H18O Molecular Weight (g/mol): 226.32 MDL Number: MFCD00275723 InChI Key: CDVRGGMPPUFSFH-UHFFFAOYSA-N Synonym: 5-Isopropyl-3,8-dimethylazulene-1-carboxaldehyde PubChem CID: 639851 IUPAC Name: 3,8-dimethyl-5-(propan-2-yl)azulene-1-carbaldehyde SMILES: CC(C)C1=CC=C(C)C2=C(C=O)C=C(C)C2=C1